BP282
Phenoxymethylpenicillin potassium
British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Penicillin V potassium salt, Phenoxymethylpenicillin potassium salt
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
penicillin & derivatives
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
[K+].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)COc3ccccc3)C([O-])=O
InChI
1S/C16H18N2O5S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)24-16)17-10(19)8-23-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1
InChI key
HCTVWSOKIJULET-LQDWTQKMSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Also used in monographs such as:
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service