Skip to Content
Merck
All Photos(1)

Key Documents

63099

Supelco

Magnesium ionophore IV - membrane A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

form

solid

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Ion selective polymer membrane for assaying magnesium in blood serum with ISE

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U.E. Spichiger et al.
Fresenius Journal of Analytical Chemistry, 341, 727-727 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service