Skip to Content
Merck
All Photos(1)

Key Documents

07410

Millipore

Carbohydrate Consumption Broth

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

according to ISO 11290-1:2017

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

beef extract, 1.0 g/L
bromo cresol purpur, 0.10 g/L
proteose peptone, 10.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

bacteria
nonselective and differential for Escherichia coli
nonselective and differential for Listeria spp.
nonselective and differential for Staphylococcus spp.
nonselective and differential for coliforms

Application

Carbohydrate consumption broth is recommended for cultivation and differentiation of Listeria species.

Preparation Note

Suspend 16.1 g in 990 ml distilled water. Heat if necessary to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 min. Aseptically add 10 ml seperately sterilized carbohydrate solution to give a final concentration of 0.5 %. Mix well and dispense in sterile test tubes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service