Key Documents
TANKPECKT
Milli-Q
Tank Connection Kit
Connector for external 30 L pure water storage reservoir
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
packaging
pkg of 1 unit
compatibility
for use with Direct-Q® UV
for use with Direct-Q®
for use with RiOs-DI® Clinical
for use with RiOs
General description
The tank connection kit connects a 30 L storage tank to the water purification system.
For use with RiOs, RiOs-DI® Clinical and Direct-Q® water purification systems.
Application
External tank connector for 30-liter stoargae tank.
Legal Information
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany
RIOS-DI is a registered trademark of Merck KGaA, Darmstadt, Germany