MIX0050LG05
Mobius® 50L Mixer Bag Assembly
Gold Mobius® 50L mixer bag
Synonym(s):
Mobius® 50 L mixer bag assy, Gold
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103801
eCl@ss:
32010600
Pricing and availability is not currently available.
Quality Level
product line
Mobius®
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service