Skip to Content
Merck
All Photos(2)

Key Documents

MAUF01005

Millipore

MultiScreen® 96-well Ultrafiltration Plate with Ultracel®-10 membrane kD

Synonym(s):

96-well centrifuge concentrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
NACRES:
NB.24

material

polypropylene plate
regenerated cellulose (RC)
silicone O-ring (medical grade)

description

For sample purification, concentration and desalting of biological solutions or protein removal

sterility

non-sterile

manufacturer/tradename

MultiScreen®

H

20.1 mm , without lid
22.0 mm , with lid

L

127.8 mm

volume

0.5 mL

width

85.5 mm

pore size

10 kDa NMWCO

storage temp.

room temp

General description

Reliability
The 96-well MultiScreen® filter plate incorporates Ultracel® -10 10,000 kDa nominal molecular weight limit regenerated cellulose ultrafiltration membrane for low-binding, high recovery results. It is designed for use with centrifugation and is compatible with standard microtiter plates, instrumentation, and liquid handling equipment.

MultiScreen® filter plate with Ultracel® -10 membrane is designed for low protein binding and high protein retention. The assay-grade plates meet stringent manufacturing release specifications and are lot tested for protein retention for >95% retention of cytochrome c (12,500 Daltons). They also offer high well-to-well uniformity of performance.

Versatile Operation
MultiScreen® filter plates with Ultracel® -10 membrane allow for processing and collection of sample volumes from 50 to 500 µL. They are compatible with a range of standard receiver 96-well microtiter plates including 300 µL, 700 µL deep well and 150 µL conical well.
The MultiScreen® 96-well Ultrafiltration Plate with Ultracel® membranes (MultiScreen Ultrafiltration Plates) are used for processing aqueous biological solutions (0.1 to 0.5 mL). The ultrafiltration plate is utilized in a centrifugal pressure mode only and is compatible with standard centrifuge microtiter plate swinging bucket rotors. It is developed to fit a 96-well microtiter receiver plate for ultrafiltrate collections. These plates have been designed specifically for protein removal before sample analysis and for sample purification, concentration, and desalting of biological solutions. 

Application

MultiScreen® 96-well Ultrafiltration Plate with Ultracel®-10 membrane kD is suitable for high-throughput protein concentration and buffer exchange.

Physical form

Colour Code: Natural

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priscilla A Moore et al.
Methods in molecular biology (Clifton, N.J.), 498, 309-314 (2008-11-07)
High-throughput protein purification is a complex, multi-step process. There are several technical challenges in the course of this process that are not experienced when purifying a single protein. Among the most challenging are the high-throughput protein concentration and buffer exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service