Skip to Content
Merck
All Photos(3)

Documents

MABN699

Sigma-Aldrich

Anti-Metadherin Antibody, clone 2F11C3

clone 2F11C3, from mouse, purified by affinity chromatography

Synonym(s):

Protein LYRIC, 3D3/LYRIC, Astrocyte elevated gene-1 protein, AEG-1, Lysine-rich CEACAM1 co-isolated protein, Metadherin, Metastasis adhesion protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F11C3, monoclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTDH(92140)

General description

Metadherin, or more commonly known as LYRIC, or AEG-1 (Astrocyte elevated gene-1) is a single pass membrane protein that plays a key role in anchorage-independent growth of immortalized melanocytes and astrocytes. Anchorage-independent division is a critical component in tumor cell expansion and metastasis. Similarly, metadherin activates the nuclear factor kappa- B (NF-kappa-B) transcription leading to expression of numerous genes involved in cell growth and expansion, and it is also involved in HIF-1alpha mediated angiogenesis in tumor growth. Finally new research demonstrates that metadherin interacts with RNA-induced silencing complex (RISC) and plays a very important role in RISC and miRNA functions as well. The protein is a key target of research because of its tumor promoting effects in lung, bone and brain cancers. Metadherin is localized to the endoplasmic reticulum and nuclear membranes in normal tissue, thus staining is perinuclear primarily but Metadherin staining can also be seen in the nucleus, at tight junctions and even at the cell surface in certain cancers. Metadherin expression is activated by TNF, and metadherin is expressed in many cells types with the highest levels in muscle-dominating organs such as skeletal muscle, heart, tongue and small intestine and in endocrine glands such as thyroid and adrenal gland. Metadherin is often over expressed in various cancers including breast, brain, prostate, and melanoma and glioblastoma multiforme. EMD-Millipore’s Anti-metadherin mouse monoclonal, clone 2F11C3 has been tested by western blot against several human cancer cell lysates, by flow cytometry against human HeLa cells, and in human paraffin-embedded human liver tissue.

Immunogen

Purified protein fragment of Human Metadherin expressed in E. coli.

Application

Detect Metadherin using this mouse monoclonal antibody, Anti-Metadherin Antibody, clone 2F11C3 validated for use in western blotting, IHC & Flow Cytometry.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Metadherin in human breast cancer tissue.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected Metadherin in HeLa cells

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Metadherin in 10 µg of K562 cell lysate.

Target description

~70 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 in PBS containing up to 0.1% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
K562 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service