Skip to Content
Merck
All Photos(1)

Key Documents

MABF2102

Sigma-Aldrich

Anti-FcRH5 Antibody, clone F56

clone F56, from mouse

Synonym(s):

Fc receptor-like protein 5, FcR-like protein 5, FcRL5, BXMAS1, Fc receptor homolog 5, Immune receptor translocation-associated protein 2, IRTA2, CD307e

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F56, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FCRL5(83416)

Related Categories

General description

Fc receptor-like protein 5 (UniProt: Q96RD9; also known as FcR-like protein 5, FcRL5, BXMAS1, Fc receptor homolog 5, FcRH5, Immune receptor translocation-associated protein 2, IRTA2, CD307e) is encoded by the FCRL5 (also known as FCRH5, IRTA2, UNQ503/PRO820) gene (Gene ID: 83416) in human. FcRH5 is a single-pass type I membrane protein that is expressed in marginal zone B-cells, immunoblasts, tonsillar germinal center centrocytes and in the intraepithelial and interfollicular regions of the tonsil. It is involved in B-cell development and differentiation in peripheral lymphoid organs and may be useful markers of B-cell stages. It is shown to regulate BCR signaling and binds aggregated IgG and engages several of its Ig-domains to bind IgG. It is shown to be overexpressed in many lymphoma cell lines and on hairy cell leukemia cells. Serum levels of soluble FcRH5 are reported to be elevated in patients with several types of B cell tumors. FcRH5 is synthesized with a signal peptide (aa 1-15), which is cleaved off in the mature form that has a long extracellular domain (aa 16-851), a short transmembrane domain (aa 852-872), and a cytoplasmic tail (aa 873-977) that contains immunoreceptor tyrosine-based inhibitor motifs (ITIM). (Ref.: Franco, A., et al. (2013). J. Immunol. 190(11); 5739-46; Ise, T., et al. (2007). Leukemia 21(1); 169-174).

Specificity

Clone F56 detects human Fc receptor-like protein 5. It targets an epitope with in the extracellular domain.

Application

Anti-FcRH5, clone F56, Cat. No. MABF2102, is a mouse monoclonal antibody that detects Fc receptor-like protein 5 and has been for use in ELISA, Flow Cytometry, Immunocytochemistry, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected FcRH5 in Immunocytochemistry applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).

Western Blotting Analysis: A representative lot detected FcRH5 in Western Blotting applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).

ELISA Analysis: A representative lot detected FcRH5 in ELISA applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).

Flow Cytometry Analysis: A representative lot detected FcRH5 in Flow Cytometry applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).

Quality

Evaluated by Flow Cytometry in SU-DHL-5 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected FcRH5 in one million SU-DHL-5 cells.

Target description

106.44 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service