Key Documents
LABCL0SE1
Milli-Q
Lab Close Kit
For use with Elix® Advantage/Reference and Milli-Q® Direct/Integral Water Purification Systems
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
packaging
pkg of 1 unit
compatibility
for use with Elix® Advantage
for use with Elix® Reference
for use with Milli-Q® Direct
for use with Milli-Q® Integral
General description
The Lab Close Kit will avoid the water purification system standstill during long inactive periods. The system when used in association with the "Lab Close" operating mode, allows you to maintain a minimum operating level.
This kit intelligently optimizes the consumption of water and electricity by your system, preventing the negative effects of nonuse, such as bacteria buildup.
Application
Limits unnecessary water consumption, while avoiding stagnation in the water flow and storage tank.
Features and Benefits
  • Minimizes water and energy usage during vacation or long inactivity periods.
  • Prevents contamination of water purification system and reservoir due to water stagnation.
  • Your system is up and running immediately when you come back.
Legal Information
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany