Skip to Content
Merck
All Photos(1)

Key Documents

8.14190

Sigma-Aldrich

Methyl isobutyrylacetate

for synthesis

Synonym(s):

Methyl isobutyrylacetate, Isobutyrylacetic acid methyl ester, Methyl 4-methyl-3-oxopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CH3)COCH2COOCH3
CAS Number:
Molecular Weight:
144.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
418-900-0
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.3 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

autoignition temp.

280 °C

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

1.29 % (v/v)

pH

4.8 (20 °C, 10 g/L in H2O)

bp

192 °C/1013 hPa

mp

-75 °C

transition temp

flash point 79 °C

solubility

43.6 g/L

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CC(=O)C(C)C

InChI

1S/C7H12O3/c1-5(2)6(8)4-7(9)10-3/h5H,4H2,1-3H3

InChI key

HNNFDXWDCFCVDM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.009 - 1.011
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service