Skip to Content
Merck
All Photos(1)

Key Documents

8.01668

Sigma-Aldrich

2-Bromopropane

for synthesis

Synonym(s):

2-Bromopropane, iso-Propyl bromide, Isopropyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHBr
CAS Number:
Molecular Weight:
122.99
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-855-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

224 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

expl. lim.

4.6 % (v/v)

bp

59-60 °C/1013 hPa

mp

-89 °C

transition temp

flash point 19 °C

density

1.31 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrC(C)C

InChI

1S/C3H7Br/c1-3(2)4/h3H,1-2H3

InChI key

NAMYKGVDVNBCFQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99,0 Area %
Density (d 20 °C/ 4 °C): 1,313 - 1,316
Identity (IR): conforms

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Repr. 1A - STOT RE 2 Inhalation

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service