Skip to Content
Merck
All Photos(1)

Key Documents

220588

Sigma-Aldrich

Chlorophenolred-β-D-galactopyranoside, Sodium Salt

≥98% (HPLC), solid

Synonym(s):

Chlorophenolred-β-D-galactopyranoside, Sodium Salt, CPRG, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21Cl2O10S · Na
Molecular Weight:
607.39
UNSPSC Code:
41116133
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Chlorophenolred-β-D-galactopyranoside, Sodium Salt,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

<0.1% chlorophenolred

color

orange-red to red

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Substrate for spectrophotometric determination of β-galactosidase activity. Exhibits about ten-fold greater sensitivity for β-galactosidase than o-NPG (Cat. No. 48712).
Substrate for the spectrophotometric determination of β-galactosidase activity. Exhibits ten-fold greater sensitivity for β-galactosidase than o-NPG (Cat. No. 48712).

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.

Other Notes

Eustice, D.C., et al. 1991. Biotechniques 11, 739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service