Skip to Content
Merck
All Photos(1)

Key Documents

1.46803

Millipore

Veg. Pept.Agar with LTHThio cont.-ICR +

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

Agency

USP 1116

Quality Level

sterility

sterile; irradiated

form

solid

shelf life

12 mo.

feature

ready-to-use

composition

Vegetable Peptone Agar with neutralizer: Lecithin, Polysorbate 80, Histidine and Sodium Thiosulfate

packaging

triple packed of

technique(s)

Rapid Sterility Testing: suitable

pH

7.1-7.5

solubility

water: soluble

application(s)

environmental
pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

15-25°C

suitability

Yeast, bacteria and molds

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service