Skip to Content
Merck
All Photos(2)

Key Documents

1.46501

Millipore

Sabouraud Dextrose Agar

Sodium thiosulphate, Tween®, Lecithin, Histidine, lid, ICR plus - lockable contact plate, plate diam. 55 mm, sterile; irradiated, triple packed of 20 or 200 plates, suitable for surface monitoring

Synonym(s):

SD Agar, SDA, SDA with neutralizer, ICRplus Contact plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

lid ICR plus - lockable contact plate
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulphate
Tween®

packaging

triple packed of 20 or 200 plates

plate diam.

55 mm

pH

5.6 ( in H2O)

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Contact Agar + LTHTh – ICR+ is designed for the determination of the total count of yeasts and molds on dry, sanitized surfaces and personnel in Clean rooms Isolators. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth. The media is supplemented with the neutralizers lecithin, polysorbate (Tween®) 80, histidine, sodium thiosulfate and pyruvate to inactivate possible residues disinfectants. In addition, the plate is supplied with a lockable lid (ICRplus; +) for safe transport after sampling without the risk of losing the lid. ICRplus plates are suitable for aerobic and anaerobic incubation by turning the lid into the close position for aerobic incubation and in the vent position for anaerobic incubation. To differentiate better SDA from TSA media, the SDA media is filled in pink dish.

Application

Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media finds its application in environmental monitoring by determining the yeast and mold count on surfaces in isolators and clean rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service