1.30110
HY-LiTE® Thermal Printer Paper Roll
Thermal roll paper for printing readings taken from HY-LiTE® 2 luminometers
Synonym(s):
Thermal printer paper, Printer paper roll, Replacement thermal roll paper
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
14111818
NACRES:
NA.74
Recommended Products
description
Thermal roll paper for printing readings taken from HY-LiTE® luminometers
compatibility
for use with HY-LITE instrument with built-in recorder (1.30100.0301)
storage temp.
no temp limit
Related Categories
General description
HY-LiTE® thermal printer paper roll is an additional accessory of HY-LiTE® 2 luminometers with built-in and external USB printer. The HY-LiTE® test system provides an easy-to-use, rapid and reliable method to assess the quality of cleaning processes in food and beverages production plants. It quantitatively measures ATP by a specific reaction with luciferase. The emitted bioluminescence is detected by the HY-LiTE® luminometer. The thermal printer paper is used for printing ATP test results taken from HY-LiTE® 2 luminometers.
Application
HY-LiTE® thermal printer paper roll is an accessory of HY-LiTE® 2 luminometers used for printing ATP test results.
Legal Information
HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service