1.10025
Total Hardness Test Strips, colorimetric
water hardness 3-21 °d, water hardness 4-26 °e, water hardness 5-37 °f, MQuant®
Synonym(s):
Total Hardness chemical test strips
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
product name
Total Hardness Test, colorimetric, water hardness 3-21 °d, water hardness 4-26 °e, water hardness 5-37 °f, MQuant®
description
semi-quantitative
Quality Level
product line
MQuant®
specific analyte(s)
calcium, magnesium
water hardness measuring range
3-21 °d
4-26 °e
5-37 °f
detection method
colorimetric
storage temp.
15-25°C
General description
Method: Colorimetric, with test strips, <3 - >4 - >7 - >14 - >21°d, <4 - >5 - >9 - >18 - >26°e, <5 - >7 - >12 - >25 - >37°f, MQuant®
The MQuant® total hardness test strips can be used to measure water hardness. The hardness (total hardness) of a given water is due to its content of salts of the alkaline earth metals calcium, magnesium, strontium, and barium (“hardening constituents”, later compounds only in traces). In the presence of a green indicator the hardness ions Ca2+ und Mg2+ react with Titriplex® (III) to form a colorless, stable complex. The hardness ions that are present in excess relative to Titriplex® (III) react with the indicator to form a red complex. The reaction zones of the test strip, which are impregnated with differing amounts of Titriplex® (III), thus assume a green to red color, depending on the hardness of the water sample. The hardness is measured semi-quantitatively by visual comparison of the reaction zones with the color rows of a color scale. For other sensitivities and graduations please consider the other MQuant® tests as well as our Reflectoquant® and Spectroquant® portfolio.
The MQuant® total hardness test strips can be used to measure water hardness. The hardness (total hardness) of a given water is due to its content of salts of the alkaline earth metals calcium, magnesium, strontium, and barium (“hardening constituents”, later compounds only in traces). In the presence of a green indicator the hardness ions Ca2+ und Mg2+ react with Titriplex® (III) to form a colorless, stable complex. The hardness ions that are present in excess relative to Titriplex® (III) react with the indicator to form a red complex. The reaction zones of the test strip, which are impregnated with differing amounts of Titriplex® (III), thus assume a green to red color, depending on the hardness of the water sample. The hardness is measured semi-quantitatively by visual comparison of the reaction zones with the color rows of a color scale. For other sensitivities and graduations please consider the other MQuant® tests as well as our Reflectoquant® and Spectroquant® portfolio.
Linkage
Replaces: 10025-1; 10025
Legal Information
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service