Skip to Content
Merck
All Photos(1)

Key Documents

1.07605

Millipore

D(-)-Ribose

for biochemistry

Synonym(s):

D(-)-Ribose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-059-4
NACRES:
NA.79

Quality Level

form

solid

pH

7 (20 °C, 10 g/L in H2O)

mp

90-95 °C

density

0.80 g/cm3 at 20 °C

bulk density

480 kg/m3

storage temp.

15-25°C

InChI

1S/C5H10O5/c6-1-2-3(7)4(8)5(9)10-2/h2-9H,1H2

InChI key

HMFHBZSHGGEWLO-UHFFFAOYSA-N

Analysis Note

Identity (IR-spectrum): passes test
Spec. rotation (α 20/D; 2 %; water): -21.0 - -18.5 °
TLC-Test: passes test
Water: ≤ 0.5 %
Heavy metals (as Pb): ≤ 0.001 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service