Skip to Content
Merck
All Photos(1)

Key Documents

1.04854

Supelco

Potassium hydrogen carbonate

for analysis EMSURE® ACS

Synonym(s):

Potassium hydrogen carbonate, Potassium bicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHKO3
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
206-059-0
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals
solubility:
362 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥99.5% (acidimetric)
99.7-100.5% dry basis (acidimetric)

form

powder or crystals

potency

>2000 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

8.4-8.6 (20 °C, 99 g/L in H2O)

solubility

362 g/L

density

2.17 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.0005%
phosphate, silicate (as SiO2): ≤0.001%
sulfate (SO42-): ≤0.001%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.001%
N: ≤0.001%
NH4+: ≤0.0005%
Na: ≤0.02%
heavy metals (as Pb): ≤0.0005%

storage temp.

15-25°C

SMILES string

[K+].[O-]C(=O)[O-].[H+]

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Application

  • Purines, pyrimidines, and imidazoles. XXXVI. Carboxylation of some 5-aminoimidazoles and related compounds, including nucleosides and nucleotides, with potassium hydrogen carbonate in aqueous solution.: This study explored the carboxylation reactions of 5-aminoimidazoles and related compounds using potassium hydrogen carbonate in aqueous solutions, highlighting its application in synthetic organic chemistry (Cusack NJ et al., 1971).

Linkage

Replaces: PX1345; PX1345-1; PX1345-30

Analysis Note

Assay (acidimetric; dried substance): 99.7 - 100.5 %
Assay (acidimetric): ≥ 99.5 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Phosphate and Silicate (as SiO₂): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.001 %
Total sulfur (as sulfate): ≤ 0.003 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
Ca (Calcium): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.001 %
Na (Sodium): ≤ 0.02 %
NH₄ (Ammonium): ≤ 0.0005 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service