Skip to Content
Merck
All Photos(1)

Key Documents

870701P

Avanti

Coenzyme A (Li salt)

Avanti Research - A Croda Brand 870701P, powder

Synonym(s):

Coenzyme A (lithium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H33Li3N7O16P3S
CAS Number:
Molecular Weight:
785.33
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 10 mg (870701P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870701P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCS)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[Li+].[Li+].[Li+]

InChI

1S/C21H36N7O16P3S.3Li/c1-21(2,16(31)19(32)24-4-3-12(29)23-5-6-48)8-41-47(38,39)44-46(36,37)40-7-11-15(43-45(33,34)35)14(30)20(42-11)28-10-27-13-17(22)25-9-26-18(13)28;;;/h9-11,14-16,20,30-31,48H,3-8H2,1-2H3,(H,23,29)(H,24,32)(H,36,37)(H,38,39)(H2,22,25,26)(H2,33,34,35);;;/q;3*+1/p-3/t11-,14-,15-,16+,20-;;;/m1.../s1

InChI key

QSCBPHBAFBVXRK-FFKZSOHASA-K

Biochem/physiol Actions

Coenzyme A is a ubiquitous cofactor, synthesized from pantothenic acid through a ATP dependent reaction.[1] It plays a vital role in tricarboxylic acid cycle,[1] metabolism of short- and long-chain fatty acids.[2] In addition, it also aids in synthesis of phospholipids and cell growth.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870701P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Leonardi et al.
EcoSal Plus, 2(2) (2007-04-01)
Pantothenate is vitamin B5 and is the key precursor for the biosynthesis of coenzyme A (CoA), a universal and essential cofactor involved in a myriad of metabolic reactions, including the synthesis of phospholipids, the synthesis and degradation of fatty acids
Roberta Leonardi et al.
Progress in lipid research, 44(2-3), 125-153 (2005-05-17)
Coenzyme A (CoA) is a ubiquitous essential cofactor that plays a central role in the metabolism of carboxylic acids, including short- and long-chain fatty acids. In the last few years, all of the genes encoding the CoA biosynthetic enzymes have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service