Skip to Content
Merck
All Photos(1)

Key Documents

860831P

Avanti

C6(6-azido) Ceramide

Avanti Research - A Croda Brand 860831P, powder

Synonym(s):

N-(6-azidohexanoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46 N4O3
CAS Number:
Molecular Weight:
438.65
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 5 mg (860831P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860831P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCN=[N+]=[N-])=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI key

ZXFVIEGVAJLSBT-PGHYDBKUSA-N

General description

C6(6-azido) Ceramide, also known as N-(6-azidohexanoyl)-D-erythro-sphingosine, is a head group modified lipid. This click reagent contains an azide group in the omega-terminal end, which can be used in a copper-catalyzed azide−alkyne click chemistry. Copper-catalyzed azide−alkyne cycloaddition reaction facilitates the connection of molecular entities of all sizes.[1]
This reagent is a modified lipid containing an omega-terminal azide. The terminal azide group can be used in a highly specific linking reaction with alkyne-containing reagents, known as click chemistry, in the presence of a copper (Cu)-containing catalyst.

Packaging

5 mL Amber Glass Screw Cap Vial (860831P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanislav I Presolski et al.
Current protocols in chemical biology, 3(4), 153-162 (2011-01-01)
The copper-catalyzed azide-alkyne cycloaddition reaction is widely used for the connection of molecular entities of all sizes. A protocol is provided here for the process with biomolecules. Ascorbate is used as reducing agent to maintain the required cuprous oxidation state.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service