Skip to Content
Merck
All Photos(2)

Key Documents

850697C

Avanti

06:0 PE

1,2-dihexanoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

PE(6:0/6:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34NO8P
CAS Number:
Molecular Weight:
411.43
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850697C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850697C

concentration

10 mg/mL (850697C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCC)=O)COC(CCCCC)=O

InChI

1S/C17H34NO8P/c1-3-5-7-9-16(19)23-13-15(26-17(20)10-8-6-4-2)14-25-27(21,22)24-12-11-18/h15H,3-14,18H2,1-2H3,(H,21,22)/t15-/m1/s1

InChI key

PELYUHWUVHDSSU-OAHLLOKOSA-N

General description

06:0 PE (1,2-dihexanoyl-sn-glycero-3-phosphoethanolamine/DHPE) is a phosphoethanolamine (PE) molecule which carries short acyl chains. It is an ethanolamine-type substrate and a water-soluble phospholipid.

Application

06:0 PE (1,2-dihexanoyl-sn-glycero-3-phosphoethanolamine/DHPE) may be used:
  • as a substrate for glycerophosphoethanolamine ethanolaminephosphodiesterase (GPE-EP) in specificity studies as a phosphoethanolamine (PE) standard in liquid chromatographic analysis of milk phospholipids
  • as a phospholipid substrate to study the kinetic parameters for the glucose transporter type 4 (Glu4) mutants
  • as calibration standard in hydrophilic interaction liquid chromatography (HILIC) and ion mobility (IM)

Biochem/physiol Actions

Phosphoethanolamine (PE) is known to possess antitumor activity and inhibitory effects on tumor progression in vivo. It also exhibits antiproliferative and proapoptotic effects. It acts as a precursor of phosphatidylcholine and phosphatidylethanolamine.

Packaging

5 mL Clear Glass Sealed Ampule (850697C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theoretical Study of Phosphoethanolamine: A Synthetic Anticancer Agent with Broad Antitumor Activity
Lorenzo VP, et al.
Journal of Chemistry (2020)
Lipidomics by HILIC-Ion Mobility-Mass Spectrometry
Ion Mobility-Mass Spectrometry, 119-132 (2020)
Peter Fagan et al.
Journal of chromatography. A, 1054(1-2), 241-249 (2004-11-24)
Two methods have been developed for the analysis of bovine milk phospholipid (PL) classes by NP-HPLC with evaporative light scattering detection. In the first method, a PVA-Sil guard column was used for the rapid determination of the major milk PL
S F Martin et al.
Biochemistry, 39(12), 3410-3415 (2000-03-22)
The phosphatidylcholine-preferring phospholipase C from Bacillus cereus (PLC(Bc)) is a 28.5 kDa enzyme with three zinc ions in its active site. The roles that a number of amino acid residues play as zinc ligands and in binding and catalysis have
Shingo Mineta et al.
Journal of bioscience and bioengineering, 119(2), 123-130 (2014-08-20)
Streptomyces sanglieri extracellularly produces a glycerophosphoethanolamine ethanolaminephosphodiesterase (GPE-EP). The gene encoding the enzyme was found to consist of a 2124-bp ORF, which codes for an N-terminal 48 residue signal peptide required for secretion and a 660 amino acid mature protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service