Skip to Content
Merck
All Photos(1)

Key Documents

330700W

Avanti

OxysterolSPLASH

Avanti Research - A Croda Brand

Synonym(s):

OxysterolSPLASH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.28
Pricing and availability is not currently available.

form

solution

packaging

package of 1 × 1 ea

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (330700W-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 700018P24(R/S)-hydroxycholesterol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700053P25-hydroxycholesterol-d6, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700059P27-hydroxycholesterol-d6, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700043P7α-hydroxycholesterol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700044P7β-hydroxycholesterol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700046P7-ketocholesterol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700112P7α-hydroxycholestenone-d7, 7α-hydroxy-4-cholesten-3-one-d7, powder 150ng/mLSDS

  • 700078P7α,25-dihydroxycholesterol-d6, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700136P7α,27-dihydroxycholesterol-d6, Avanti Research - A Croda Brand 700136P, powder 150ng/mLSDS

  • 700194P7α-Hydroxy-3-oxocholest-4-enoic acid-d3, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700042P4β-hydroxycholesterol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700052P22(R)-hydroxycholesterol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

  • 700055P5α,6β-dihydroxycholestanol-d7, Avanti Research - A Croda Brand 150ng/mLSDS

See All (13)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service