Z743741
ART® Low Retention Barrier Tip
ART 20P, size 200 μL, racked, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
About This Item
racked
Recommended Products
sterility
sterile
feature
ART 20P
racked
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 2149P-05
technique(s)
PCR: suitable
size
200 μL
volume
200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- ART® low retention barrier tips are designed to maximize sample recovery.
- Tip′s hydrophobic inner surface prevents sample loss, facilitates sample accuracy and reproducibility.
- Clear tips provide sample clarity.
- Self-sealing technology prevents cross-contamination.
- Certified RNase, DNase, ATP, endotoxin, and pyrogen free.
Features and Benefits
- ART low retention barrier tips ensure complete protection against aerosols and liquid contamination.
- The tips are free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
- Stackable barrier tip packaging enables tips to pass through offset holes in the lower insert.
- Blister sleeve guide rails facilitate smooth transfer of tip inserts into the rack.
- An outer sleeve shields tips from external contamination during the refilling of inserts.
- The tip packaging option helps reduce plastic waste.
- The rigid insert design ensures reliable tip loading.
- Lot number and description are printed on every insert for easy tracking and identification.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service