Skip to Content
Merck
All Photos(2)

Key Documents

Z676578

ART® low retention barrier tips

Tip design ART 10 REACH, Hinged rack

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
10 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
endotoxin free
Hinged rack
Tip design ART 10 REACH
packaging:
pack of 4800 ea (50 hinged racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2140-05-HR

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
endotoxin free
Hinged rack
Tip design ART 10 REACH

packaging

pack of 4800 ea (50 hinged racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2140-05-HR

technique(s)

PCR: suitable

maximum volume

10 μL

volume

10 μL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

ART® low retention barrier tips in hinged racks provide superior sealing capabilities when compared to standard filter tips. The tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks, and the covers can be used in either hinged or lift-off style. The ART barrier comes in eco-friendly packaging. The sterile reload system includes blister tubs made from recycled plastic and new hinged racks. You can conveniently insert the ART barrier tips into the hinged racks using the push-down blister tub, creating stackable racks that save space and provide one-handed operation. You can easily convert to the ART Reload System with the part numbers for empty hinged racks.

Application

ART® low retention barrier tips are suitable for:
  • Liquid handling
  • Molecular biology
  • PCR
  • Biotechnology
  • Pipetting radioactive samples
  • Nucleic amplification procedures

Features and Benefits

  • Hinged racks that can be stacked help conserve lab space.
  • DNase, RNase, DNA, ATP, PCR inhibitor, and endotoxin free.
  • The locking clasp ensures the remaining tips remain sterile, preventing contamination, and offers convenient one-handed operation.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service