Merck
All Photos(6)
Key Documents
Z509140
Sigma-Aldrich
Scissor-handle forceps
Occulating, straight, L 7 in.
About This Item
UNSPSC Code:
41122405
NACRES:
NB.35
Pricing and availability is not currently available.
label
D
packaging
pack of 1 ea
L
7 in.
General description
stainless steel hemostatic forcep with smooth jaw and box-lock joint
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2