Skip to Content
Merck
All Photos(1)

Key Documents

QBD11812

Sigma-Aldrich

DBCO-dPEG ®12-carboxyfluorescein

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H79N3O20
Molecular Weight:
1234.34
UNSPSC Code:
51171641
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reagent type: fluorescent-labelling reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(C)NCCOCCC(NCCC(N1C(C=CC=C2)=C2C#CC(C=CC=C3)=C3C1)=O)=O.O=C4C=CC5=C(C6=CC=CC=C6C(O)=O)C7=CC=C(O)C=C7OC5=C4

InChI

1S/C25H27N3O4.C20H12O5/c1-19(29)26-15-17-32-16-13-24(30)27-14-12-25(31)28-18-22-8-3-2-6-20(22)10-11-21-7-4-5-9-23(21)28;21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24/h2-9H,12-18H2,1H3,(H,26,29)(H,27,30);1-10,21H,(H,23,24)

InChI key

PMSFOKNHIHSIKS-UHFFFAOYSA-N

Application

DBCO-dPEG®12-carboxyfluorescein is a versatile fluorescent reagent used for labeling peptides, proteins, and other molecules using strain-promoted azide-alkyne cycloaddition (SPAAC).

Features and Benefits

DBCO-dPEG®₁₂-carboxyfluorescein is a fluorescent dye with a dPEG spacer attached to a DBCO which can react with azide through copper-free "click" chemistry.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service