OCNQAK540
SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots
fluorescence λem 540 nm, for 4 conjugation
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
fluorescence
λem 540 nm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Components:
- QSA
- Sulfo-SMCC
- Traut′s Reagent
- Coupling Buffer
- Storage Buffer
- Quenching Buffer
- Tubes 4 x 1.7 mL
Legal Information
Product of Ocean Nanotech, LLC.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service