GF60137917
Copper-beryllium alloy, Cu98Be2
foil, 25 x 25mm, 0.02mm thickness, half hard, light tight (lt)
Synonym(s):
CU070276
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
31152304
NACRES:
NA.23
description
light tight (lt)
form
foil (half hard)
manufacturer/tradename
Goodfellow 601-379-17
size × thickness
25 mm × 25 mm × 0.02 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service