GF03846124
Copper-beryllium alloy, Cu98Be2
foil, 0.5m coil, .3mm coil width, 0.15mm thickness, half hard
Synonym(s):
CU070361
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
31152304
NACRES:
NA.23
Recommended Products
description
temper (half hard)
form
foil (coil)
manufacturer/tradename
Goodfellow 038-461-24
L × W × thickness
0.5 m × 0.3 mm × 0.15 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service