Skip to Content
Merck
All Photos(1)

Key Documents

BR759710

BRAND® disposal bag

PA

Synonym(s):

Bags, Disposal Bags

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

material

PA

manufacturer/tradename

BRAND 759710

L × W

300 mm × 200 mm

Looking for similar products? Visit Product Comparison Guide

General description

Clear polyamide laboratory disposal bags are designed to provide a reliable and durable solution for the safe containment and disposal of laboratory waste. Constructed from polyamide, also known as nylon, these bags offer exceptional strength and resistance to tearing and punctures. The transparent design allows for easy identification and visual inspection of the bag′s contents. These bags are suitable for containing non-sharp laboratory items, offering a secure and organized means of waste disposal.

Application

Waste disposal

Features and Benefits

  • Made of polyamide
  • Autoclavable
  • Sharp objects, such as needles, should never be placed in this bag

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service