BR37413
BRAND® BLAUBRAND® volumetric flask
polypropylene stopper, amber borosilicate glass 3.3 flask, capacity 25 mL, accuracy, 0.06 mL
Synonym(s):
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
About This Item
Recommended Products
material
amber borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint ST/NS12/21
description
calibrated to contain (TC, IN)
feature
accuracy 0.06 mL
class A DIN EN ISO 1042. Calibrated to contain (TC, In)
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37413
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service