Skip to Content
Merck
All Photos(2)

Key Documents

901275

Sigma-Aldrich

Sodium ethyl sulfate

≥98%

Synonym(s):

Sodium sulfovinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5NaO4S
CAS Number:
Molecular Weight:
148.11
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder or crystals

SMILES string

[Na+].CCOS([O-])(=O)=O

InChI

1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1

InChI key

RRLOOYQHUHGIRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Building block for chemical synthesis. Sodium ethyl sulfate may also be used in the preparation of sodium ethane sulfonate by heating it in concentrated sodium sulfite aqueous solution. This product was previously listed as CDS000593.
Sodium ethyl sulfate can be converted to nitroethane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of nitroalkanes.
Aitken R A and Aitken K M
Sci. Synth., 41, 9-258 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service