Skip to Content
Merck
All Photos(2)

Key Documents

750816

Sigma-Aldrich

N-4-Boc-aminocyclohexanone

95%

Synonym(s):

(4-Oxocyclohexyl)carbamicacid tert-butyl ester, 4-(tert-Butoxycarbonylamino)cyclohexanone, tert-Butyl 4-oxocyclohexylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

114-118 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NC1CCC(=O)CC1

InChI

1S/C11H19NO3/c1-11(2,3)15-10(14)12-8-4-6-9(13)7-5-8/h8H,4-7H2,1-3H3,(H,12,14)

InChI key

WYVFPGFWUKBXPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service