Skip to Content
Merck
All Photos(1)

Key Documents

700428

Sigma-Aldrich

N,N-Dimethylformamide-d7

≥99.5 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

DMF-d7, Heptadeutero-N,N-dimethylformamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCON(CD3)2
CAS Number:
Molecular Weight:
80.14
Beilstein:
1908468
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99.5 atom % D

Assay

≥99% (CP)

form

solution

contains

0.03 % (v/v) TMS

technique(s)

NMR: suitable

impurities

≤0.05% water
water

refractive index

n20/D 1.428 (lit.)

bp

153 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

mass shift

M+7

SMILES string

[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3,3D

InChI key

ZMXDDKWLCZADIW-YYWVXINBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide-d7 (DMF-d7) is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. Pulsed neutron diffraction measurements of different concentrations of molar lithium chloride (LiCl) solutions in DMF-d7 have been carried out to investigate the chloride ion solvation.

Application

N,N-Dimethylformamide-d7 may be used to analyze the degree of acrylation by proton NMR during the preparation of polyethylene glycol diacrylate (PEG-DA) from poly(ethylene glycol) (PEG).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Furse et al.
FEBS letters, 589(19 Pt B), 2726-2730 (2015-08-15)
This paper describes a study of the phospholipid profile of Escherichia coli MG1655 cultures at the B and D periods of the cell cycle. The results indicate that the phosphatidyl glycerol fraction grows relatively rapidly and that the size of
Neutron diffraction study on chloride ion solvation in water, methanol, and N,N-dimethylformamide.
Yamagami M, et al.
J. Chem. Phys. , 103(18), 8174-8178 (1995)
Seung Bin Baek et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(46), 14156-14161 (2015-11-19)
Crystallographic observation of adsorbed gas molecules is a highly difficult task due to their rapid motion. Here, we report the in situ single-crystal and synchrotron powder X-ray observations of reversible CO2 sorption processes in an apparently nonporous organic crystal under
Małgorzata Fabijańska et al.
Dalton transactions (Cambridge, England : 2003), 44(3), 938-947 (2014-08-12)
This paper describes the synthesis of trans-bis-(3-aminoflavone)dichloridoplatinum(ii) (trans-Pt(3-af)2Cl2; TCAP) for use as a potential anticancer compound, and the evaluation of its structure by elemental and spectral analyses, and X-ray crystallography. The complex demonstrated a significant cytotoxic effect against human and
Sean E Doris et al.
Journal of the American Chemical Society, 136(44), 15702-15710 (2014-10-11)
Cationic naked nanocrystals (NCs) are useful building units for assembling hierarchical mesostructured materials. Until now, their preparation required strongly electrophilic reagents that irreversibly sever bonds between native organic ligands and the NC surface. Colloidal instabilities can occur during ligand stripping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service