Skip to Content
Merck
All Photos(1)

Key Documents

518883

Sigma-Aldrich

Dichlorodiphenoxymethane

Synonym(s):

Phosgene diphenyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2CCl2
CAS Number:
Molecular Weight:
269.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

39-44 °C (lit.)

SMILES string

ClC(Cl)(Oc1ccccc1)Oc2ccccc2

InChI

1S/C13H10Cl2O2/c14-13(15,16-11-7-3-1-4-8-11)17-12-9-5-2-6-10-12/h1-10H

InChI key

TVIUSKXXXZSVJU-UHFFFAOYSA-N

Application

Reactant for:
  • Cyclocondensation of aminophenols and amines
  • Preparation of LIM-Kinase 2 inhibitors for the treatment of ocular hypertension

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and mass spectral fragmentations of new spiro heterocycles.
Rahimizadeh M, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 18(6), 689-693 (2007)
The Reactions of Methyl-and n-Butyllithium with Dichloromethyl Methyl Ether and Dichlorodiphenoxymethane.
McDonald RN and Krueger.
The Journal of Organic Chemistry, 30(12), 4372-4375 (1965)
Iodine-Catalyzed Synthesis of Spiroorthocarbonates under Neutral Conditions.
Rahimizadeh M, et al.
ChemInform, 41(1), i-i (2010)
The synthesis of sulfur-containing spiro orthocarbonates.
Bromley MK, et al.
Australian Journal of Chemistry, 52(7), 657-662 (1999)
Polyorthocarbonates.
Takekoshi T.
Journal of Polymer Science Part A: Polymer Chemistry, 10(12), 3509-3518 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service