Skip to Content
Merck
All Photos(1)

Key Documents

461199

Sigma-Aldrich

2,4,7,9-Tetramethyl-5-decyne-4,7-diol ethoxylate

average Mn 670

Synonym(s):

2,4,7,9-Tetramethyl-5-decyne-4,7-diol-ethylene oxide adduct, 2,4,7,9-tetramethyl-5-decyne-4,7-diol, Acetylenol EL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)[(-OCH2CH2-)mOH]C≡CC(CH3)[(-OCH2CH2-)nOH]CH2CH(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mn 670

Quality Level

refractive index

n20/D 1.468

pH

6-8

surface tension

47 dyn/cm, 0.01 wt. % in H2O

bp

>121 °C (lit.)

transition temp

cloud point 63 °C
pour point 7 °C

solubility

carbon tetrachloride, xylene and ethylene glycol: soluble

density

1.04 g/mL at 25 °C

HLB

13

SMILES string

OCCO.CC(C)CC(C)(O)C#CC(C)(O)CC(C)C

InChI

1S/C14H26O2.C2H6O2/c1-11(2)9-13(5,15)7-8-14(6,16)10-12(3)4;3-1-2-4/h11-12,15-16H,9-10H2,1-6H3;3-4H,1-2H2

InChI key

SUHUKEQAOUOUJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Surfactant.
Reduces surface tension; wetting agent, defoamer, and emulsifier for emulsion polymerization.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service