454923
3-Chloro-2-hydroxypropyl methacrylate
Synonym(s):
(3-Chloro-2-hydroxypropyl) 2-methylprop-2-enoate, 2-Hydroxy-3-chloropropyl methacrylate, 3-Chloro-2-hydroxypropyl methacrylate
About This Item
Recommended Products
contains
250 ppm monomethyl ether hydroquinone as inhibitor
Quality Level
refractive index
n20/D 1.475 (lit.)
bp
95 °C/2 mmHg (lit.)
density
1.19 g/mL at 25 °C (lit.)
SMILES string
CC(=C)C(=O)OCC(O)CCl
InChI
1S/C7H11ClO3/c1-5(2)7(10)11-4-6(9)3-8/h6,9H,1,3-4H2,2H3
InChI key
DDKMFQGAZVMXQV-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service