341762
1,12-Diisocyanatododecane
97%
Synonym(s):
1,12-Diisocyanatododecane, 1,12-Dodecandiisocyanate, Dodecamethylene diisocyanate
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.459 (lit.)
bp
168-169 °C/3 mmHg (lit.)
density
0.94 g/mL at 25 °C (lit.)
SMILES string
O=C=NCCCCCCCCCCCCN=C=O
InChI
1S/C14H24N2O2/c17-13-15-11-9-7-5-3-1-2-4-6-8-10-12-16-14-18/h1-12H2
InChI key
GFNDFCFPJQPVQL-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service