296686
(S)-(+)-3-Hydroxytetrahydrofuran
99%
Synonym(s):
(S)-(+)-Tetrahydro-3-furanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
4652609
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
optical activity
[α]20/D +17.5°, c = 2.4 in methanol
optical purity
ee: 99% (GLC)
refractive index
n20/D 1.45 (lit.)
bp
80 °C/15 mmHg (lit.)
density
1.103 g/mL at 25 °C (lit.)
SMILES string
O[C@H]1CCOC1
InChI
1S/C4H8O2/c5-4-1-2-6-3-4/h4-5H,1-3H2/t4-/m0/s1
InChI key
XDPCNPCKDGQBAN-BYPYZUCNSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
174.2 °F - closed cup
Flash Point(C)
79 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service