Skip to Content
Merck
All Photos(1)

Key Documents

217867

Sigma-Aldrich

2-Bromo-5-(trifluoromethyl)aniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CF3)NH2
CAS Number:
Molecular Weight:
240.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.522 (lit.)

density

1.675 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Nc1cc(ccc1Br)C(F)(F)F

InChI

1S/C7H5BrF3N/c8-5-2-1-4(3-6(5)12)7(9,10)11/h1-3H,12H2

InChI key

PZDVFXUBTKPFSG-UHFFFAOYSA-N

Application

2-Bromo-5-(trifluoromethyl)aniline was used in the synthesis and biochemical evaluation of series of inhibitors of hepatitis C virus (HCV) NS3 protease[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Rönn et al.
Bioorganic & medicinal chemistry, 16(6), 2955-2967 (2008-01-16)
Inhibition of the hepatitis C virus (HCV) NS3 protease has emerged as an attractive approach to defeat the global hepatitis C epidemic. In this work, we present the synthesis and biochemical evaluation of HCV NS3 protease inhibitors comprising a non-natural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service