Merck
  • Leflunomide inhibition of BK virus replication in renal tubular epithelial cells.
Leflunomide inhibition of BK virus replication in renal tubular epithelial cells.
Journal of virology (2009-12-04)
Eva Bernhoff, Garth D Tylden, Lars J Kjerpeseth, Tore J Gutteberg, Hans H Hirsch, Christine H Rinaldo
ABSTRACT
The immunomodulatory drug leflunomide is frequently used for treating polyomavirus-associated nephropathy, yet its antiviral mechanism is unclear. We characterized the effects of the active leflunomide metabolite A771726 (LEF-A) on the polyomavirus BK (BKV) life cycle in human renal tubular epithelial cells. LEF-A at 10 microg/ml reduced the extracellular BKV load by 90% (IC(90)) but with significant host cytostatic effects. BKV genome replication, late protein expression, and virion assembly and release were inhibited with visible disruption of the nuclear replication architecture. Both host cell and antiviral effects were largely reversed by uridine addition, implicating nonspecific pyrimidine depletion as the major anti-BKV mechanism of leflunomide.
Product Description
USP
Leflunomide Related Compound B, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.