Merck
  • Circulating uromodulin inhibits systemic oxidative stress by inactivating the TRPM2 channel.
Circulating uromodulin inhibits systemic oxidative stress by inactivating the TRPM2 channel.
Science translational medicine (2019-10-04)
Kaice A LaFavers, Etienne Macedo, Pranav S Garimella, Camila Lima, Shehnaz Khan, Jered Myslinski, Jeanette McClintick, Frank A Witzmann, Seth Winfree, Carrie L Phillips, Takashi Hato, Pierre C Dagher, Xue-Ru Wu, Tarek M El-Achkar, Radmila Micanovic
ABSTRACT
High serum concentrations of kidney-derived protein uromodulin [Tamm-Horsfall protein (THP)] have recently been shown to be independently associated with low mortality in both older adults and cardiac patients, but the underlying mechanism remains unclear. Here, we show that THP inhibits the generation of reactive oxygen species (ROS) both in the kidney and systemically. Consistent with this experimental data, the concentration of circulating THP in patients with surgery-induced acute kidney injury (AKI) correlated with systemic oxidative damage. THP in the serum dropped after AKI and was associated with an increase in systemic ROS. The increase in oxidant injury correlated with postsurgical mortality and need for dialysis. Mechanistically, THP inhibited the activation of the transient receptor potential cation channel, subfamily M, member 2 (TRPM2) channel. Furthermore, inhibition of TRPM2 in vivo in a mouse model mitigated the systemic increase in ROS during AKI and THP deficiency. Our results suggest that THP is a key regulator of systemic oxidative stress by suppressing TRPM2 activity, and our findings might help explain how circulating THP deficiency is linked with poor outcomes and increased mortality.
Sigma-Aldrich
DAPI, for nucleic acid staining
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rac1 Activation Assay Kit, Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Rac1-GTP from cell lysates & detection by a Rac1 specific monoclonal antibody.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Human UMOD / Uromodulin ELISA Kit