
Moxifloxacin Related Compound D

Synonym(s):
1-Cyclopropyl-8-fluoro-6-methoxy-1,4-dihydro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid
Empirical Formula (Hill Notation):
C21H24FN3O4
CAS Number:
Molecular Weight:
401.43
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing