
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing