
5(6)-Carboxytetramethylrhodamine

Synonym(s):
5(6)-Carboxytetramethylrhodamine
Empirical Formula (Hill Notation):
C25H22N2O5
CAS Number:
Molecular Weight:
430.45
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Showing 1-2 of 2