Skip to Content
Merck
All Photos(1)

Key Documents

8.51030

Sigma-Aldrich

5(6)-Carboxytetramethylrhodamine

Novabiochem®

Synonym(s):

5(6)-Carboxytetramethylrhodamine, 5(6)-TAMRA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22N2O5
CAS Number:
Molecular Weight:
430.45
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥97% (TLC)

form

powder

reaction suitability

reagent type: fluorescent-labelling reagent

manufacturer/tradename

Novabiochem®

mp

>300 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl.Cl.N(C)(C)c5cc6[o+]c7c(c(c6cc5)c8c(cccc8C(=O)O)C(=O)[O-])ccc(c7)N(C)C.N(C)(C)c1cc2[o+]c3c(c(c2cc1)c4c(ccc(c4)C(=O)O)C(=O)[O-])ccc(c3)N(C)C

InChI

1S/2C25H22N2O5.2ClH/c1-26(2)15-6-9-18-21(12-15)32-22-13-16(27(3)4)7-10-19(22)23(18)20-11-14(24(28)29)5-8-17(20)25(30)31;1-26(2)14-8-10-16-20(12-14)32-21-13-15(27(3)4)9-11-17(21)22(16)23-18(24(28)29)6-5-7-19(23)25(30)31;;/h2*5-13H,1-4H3,(H-,28,29,30,31);2*1H

InChI key

HMIPWWWHTRTUAD-UHFFFAOYSA-N

General description

Novabiochem®′s 5(6)-carboxytetramethylrhodamine (TAMRA; λem555 nm, λex580 nm) is a useful tool for preparing fluorescently-labeled peptides. It is supplied as a mixture of 5-and 6-isomers, making it cost effective for those applications which do not require a single isomer dye.


Associated Protocols and Technical Articles
Chromogenic Labeling of Synthetic Peptides

Linkage

Replaces: 01-63-0134

Analysis Note

Colour (visual): dark red to grey-purple
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(011C)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service