Skip to Content
Merck
All Photos(1)

Key Documents

57527

Supelco

Docosanoic-1,2,3,4-13C4 acid

analytical standard

Synonym(s):

Behenic acid-1,2,3,4-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C18H44O2
Molecular Weight:
344.55
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.13
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

storage temp.

2-8°C

SMILES string

O=[13C](O)[13CH2][13CH2][13CH2]CCCCCCCCCCCCCCCCCC

InChI

1S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)/i19+1,20+1,21+1,22+1

InChI key

UKMSUNONTOPOIO-ALVDIJLPSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service