The Micro Connectors (67400-U) used with ATIS extraction glassware 28523-U are equipped with phenolic caps containing a Teflon inner part for the passage of the sample. While no specific pressure rating is provided for these connectors, both the Micro-Connector and Ground-Glass versions can be used at =/< 25" of water at a 125ml/minute flow rate. it is recommended to consider using the 19/22 ground-glass version as a replacement for the Micro Connector (64700-U). Additionally, manuals for the Purge and Trap/Humidifier Module Accessory are available upon request.
28523-U
ATIS Extraction Glassware with Micro Connector
About This Item
Recommended Products
fitting
connector (20/20)
application(s)
air monitoring
environmental
industrial hygiene
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Chromatograms
suitable for GC, application for air monitoringsuitable for GC, application for air monitoringsuitable for GC, application for air monitoringsuitable for GC, application for air monitoringShow More-
Provide details about the materials used in the joint of Micro Connector 28523-U and the pressures it can accept? Additionally, having access to a user manual for this product would be helpful in determining if it is suitable as per the needs.
1 answer-
Helpful?
-