Skip to Content
Merck
All Photos(1)

Key Documents

21123

Supelco

GC Stationary Phase

phase Triton X-100, bottle of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

description

non-ionic

form

liquid

packaging

bottle of 50 g

parameter

temp. range at 0-200 °C

technique(s)

gas chromatography (GC): suitable

matrix active group

Triton X-100 phase

solubility

acetone: soluble

column type

packed GC

Looking for similar products? Visit Product Comparison Guide

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered before choosing a stationary phase. Triton X-100 is a non-ionic alkylphenol ethoxylate surfactant.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Aranda et al.
Journal of chromatography. A, 888(1-2), 35-41 (2000-08-19)
Two polycrystalline graphites (pencil lead and glassy carbon) were used as sorbents for solid-phase microextraction of a nonionic alkylphenol ethoxylate surfactant (Triton X-100). Analyses were performed by reversed-phase HPLC-fluorescence detection. The presence of the benzene ring in the congeners of
Jianyu He et al.
Annals of translational medicine, 8(21), 1400-1400 (2020-12-15)
In liver tissue engineering, co-culturing hepatocytes with typical non-parenchymal hepatic cells to form cell aggregates is available to mimic the in vivo microenvironment and promote cell biological functions. With a modular assembly approach, endothelialized hepatic cell aggregates can be packed
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service