Key Documents
Z370819
Scienceware® replicator, 96-well
polycarbonate
Synonym(s):
plate replicator
About This Item
UNSPSC Code:
41122102
NACRES:
NB.22
Pricing and availability is not currently available.
material
polycarbonate
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37876-0002
General description
Perform tedious 96 well blotting procedures with one simple motion. The 96 "open ended" pipette tips in this unique product fit all types of plates from flat, vee or round bottom plates to 0.2 mL thin walled PCR plates and tubes. The replicator fits 96 well plates of any standard configuration, and will pick up about 10 μL from each well. Then deliver the transfer to a hybridization membrane or an agar surface. Suitable for replication of rDNA libraries, colony hybridization techniques, phage typing, etc.
  • Autoclavable
  • Polypropolyene
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)